Anti-Eng Recombinant Antibody [Clone: MJ7/18]
Catalog # PRSI24-442
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:endoglin
- Antigen symbol:Eng
- Clonality:Recombinant
- Clone:MJ7/18
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:13805
- Antigen synonyms:Endoglin|Cell surface MJ7/18 antigen
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:MJ7/18 was prepared by immunizing rats with inflamed mouse skin tissue. The antibody was selected immunohistologically for reactivity with endothelial cells
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: endoglin
Clonality: Recombinant
Clone: MJ7/18
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse