Anti-CD40 Recombinant Antibody [Clone: G28.5]
Catalog # PRSI24-132
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD40 molecule
- Antigen symbol:CD40
- Clonality:Recombinant
- Clone:G28.5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:958
- Antigen synonyms:Tumor necrosis factor receptor superfamily member 5|B-cell surface antigen CD40|CD154R|TNFRS5|Bp50|CD40L receptor CDw40
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human erythrocyte-rosette-negative tonsillar lymphocytes
- Tested applications:Activation
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD40 molecule
Clonality: Recombinant
Clone: G28.5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human