Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-NIP Recombinant Antibody [Clone: B1-8]
  PRSI24-091
New Product
Anti-NIP Recombinant Antibody [Clone: B1-8]
  PRSI24-091
New Product

 

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Clone:
    B1-8
  • Conjugation:
    Unconjugated
  • Host:
  • Isotype:
    IgG1 lambda
  • Reactivity:
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    NIP
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    hapten-protein conjugated NP12-chicken gammaglobulin (CG)
  • Tested applications:
    Negative control
  • Pack type:
    Vial
  • Pk:
    0,2 mg

 

 

Type: Primary
Antigen: NIP
Clonality: Recombinant
Clone: B1-8
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 lambda
Reactivity: