Anti-MARCO Recombinant Antibody [Clone: PLK1]
Catalog # PRSI24-701
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:macrophage receptor with collagenous structure
- Antigen symbol:MARCO
- Clonality:Recombinant
- Clone:PLK1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:8685
- Antigen synonyms:PLK 1|macrophage receptor MARCO|SCARA2|scavenger receptor class A member 2|macrophage receptor with collagenous structure|PLK-1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The antibody was raised by immunising BALB/c mice with human alveolar macrophages (AMs) and subsequent creation of hybridomas
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: macrophage receptor with collagenous structure
Clonality: Recombinant
Clone: PLK1
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human