Bromocresol purple sodium salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate
CAS Number:62625-30-3
:
Formula:C₂₁H₁₅Br₂NaO₅S
MDL Number:MFCD00148896
Molecular Weight:562.21 g/mol
Melting Pt:255
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:263-655-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together