KN-93 (water soluble) ≥98% (by 1H-NMR)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: (E)-N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-N-(2-hydroxyethyl)-4-methoxybenzenesulphonamide phosphate, (E)-KN-93 phosphate
CAS Number:1188890-41-6 | 139298-40-1 (free base)
:
Formula:C₂₆H₂₉ClN₂O₄S·H₃PO₄
MDL Number:MFCD03788201
Molecular Weight:501.0·98.0 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together