PD-146176 ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: 6,11-Dihydrothiochromeno[4,3-b]indole, PD146176
CAS Number:4079-26-9
:
Formula:C₁₅H₁₁NS
MDL Number:MFCD05664738
Molecular Weight:237.3 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together