8-Bromo-cAMP sodium salt ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: 8- Bromoadenosine- 3', 5'- cyclic monophosphate, 8-Br-cAMP, 8-Br-Camp sodium salt, 8-Bromoadenosine 3',5'-cyclic monophosphate sodium salt, 8-Bromo-cAMP, 8- Bromoadenosine- 3', 5'-monophosphate
CAS Number:76939-46-3
:
Formula:C₁₀H₁₀BrN₅NaO₆P
MDL Number:MFCD00005844
Molecular Weight:430.08 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity:≥98% (HPLC)
- Identity:Determined by ¹H-NMR, ³¹P-NMR, ESI-MS and UV
Specification Test Results
Frequently Bought Together