β-Lapachone ≥98%
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: 2,2-Dimethyl-3,4-dihydro-2H-benzo[h]chromene-5,6-dione
CAS Number:4707-32-8
:
Formula:C₁₅H₁₄O₃
MDL Number:MFCD01712233
Molecular Weight:242.274 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together