8-(4-Chlorophenylthio)adenosine-3',5'-cyclic monophosphate sodium salt (synthetic) ≥96% (by HPLC)
New Product
: 8-CPT cAMP
:93882-12-3
:
:C₁₆H₁₄ClN₅NaO₆PS
:MFCD00057683
:493.791 g/mol
:Freezer
- Environmentally Preferable:
- Source:Synthetic
- Purity:≥96%
Frequently Bought Together