4-Amino-3-methylbenzonitrile 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Cyano-2-methylaniline
CAS Number:78881-21-7
:
Formula:C₈H₈N₂
MDL Number:MFCD02093969
Boiling Pt:292
Molecular Weight:132.17
Melting Pt:93…94 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1 III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together