L-Alanine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
CAS Number:56-41-7
:
Formula:C₃H₇NO₂
MDL Number:MFCD00064410
Boiling Pt:189
Melting Pt:79
Density:1.4 g/cm³ (20 °C)
Molecular Weight:89.09 g/mol
Merck Index:13,00203
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-273-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together