Xylitol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
CAS Number:87-99-0
:
Formula:C₅H₁₂O₅
MDL Number:MFCD00064292
Boiling Pt:495
Melting Pt:92…96 °C
Density:1.515
Molecular Weight:152.15 g/mol
Merck Index:13,10140
Storage Temperature:Ambient
EINECS:201-788-0
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together