4-Bromo-2,6-xylenol, Sigma-Aldrich®
Catalog # SIAL196371-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Bromo-2,6-dimethylphenol, 5-Bromo-2-hydroxy-3-xylene
:
:201.06 g/mol
EINECS:219-153-1
CAS Number:2374-05-2
Formula:C₈H₉BrO
MDL Number:MFCD00002314
Boiling Pt:259
Melting Pt:79
:M-Clarity™ quality level = MQ100