4-Bromo-3,5-xylenol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Bromo-3,5-dimethylphenol, 2-Bromo-5-hydroxy-3-xylene
CAS Number:7463-51-6
:
Formula:C₈H₉BrO
MDL Number:MFCD00002315
Boiling Pt:259
Molecular Weight:201.06 g/mol
Melting Pt:113…116 °C
EINECS:231-255-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to light pink Powder or crystals
- Infrared spectrum:Conforms
- Melting point:113 to 116 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together