Hexyl formate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Formic acid hexyl ester, n-Hexyl formate
CAS Number:629-33-4
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00046146
Boiling Pt:155
Melting Pt:–62.6 °C
Density:0.875 g/cm³ (20 °C)
Molecular Weight:130.19 g/mol
EINECS:211-087-1
ADR:3 II
:M-Clarity™ quality level = MQ300
UN:3272
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together