Neutral red, powder, Bioreagent cell culture reagent, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
CAS Number:553-24-2
:
Formula:C₁₅H₁₇ClN₄
MDL Number:MFCD00012651
Molecular Weight:288.78 g/mol
Melting Pt:290 (dec., lit.)
Merck Index:12,06571
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:209-035-8
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour):Brown to Black
may have red or green sheen - Appearance (Form):Powder
- Solubility (Colour)
10 mg/ml, H₂O:Dark Red - Infrared spectrum:Conforms to Structure
- Dye Content
(Calculated based on UV spectral assay):≥90 % - Cell Culture Test:Pass
- Recommended Retest Period:4 years
Specification Test Results
Frequently Bought Together