4,4'-Dihydroxybenzophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Bis(4-hydroxyphenyl) ketone
CAS Number:611-99-4
:
Formula:C₁₃H₁₀O₃
MDL Number:MFCD00002358
Boiling Pt:375
Melting Pt:214…217 °C
Density:1.336 g/cm³ (20 °C)
Molecular Weight:214.22 g/mol
:M-Clarity™ quality level = MQ100
EINECS:210-288-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together