Order Entry
United Kingdom
ContactUsLinkComponent
Nitric acid 90% fuming fuming ≥90% ACS, Sigma-Aldrich®
Nitric acid 90% fuming fuming ≥90% ACS, Sigma-Aldrich®
Catalog # SIAL695041-500ML
Supplier:  Merck
CAS Number:  
Nitric acid 90% fuming fuming ≥90% ACS, Sigma-Aldrich®
Catalog # SIAL695041-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Nitric acid solution
 :
 :63.01 g/mol
EINECS:231-714-2
UN:2032
REACH:01-2119487297-23
 :GHS03
 :GHS05
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:87
Melting Pt:–42 °C
Density:1.5 g/cm³ (20 °C)
ADR:8 I
Merck Index:13,06610