2-Fluoro-4-(trifluoromethyl)benzaldehyde, Sigma-Aldrich®
Catalog # SIAL529214-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:192.11 g/mol
UN:1987
CAS Number:89763-93-9
Formula:C₈H₄F₄O
MDL Number:MFCD00061310
Boiling Pt:118…119 °C (1013 hPa)
Density:1.41 g/cm³ (20 °C)
ADR:3 III
:M-Clarity™ quality level = MQ100