cis-4-Decenal, Sigma-Aldrich®
Catalog # SIAL375624-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (Z)-4-Decenal
:
:154.25 g/mol
EINECS:244-514-5
UN:3334
:GHS07
CAS Number:21662-09-9
Formula:C₁₀H₁₈O
MDL Number:MFCD00007024
Boiling Pt:78
Density:0.8569 g/cm³ (20 °C)
ADR:9
:M-Clarity™ quality level = MQ100