Ethyl dichloroacetate, Sigma-Aldrich®
SIAL35820-100ML
: Merck
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
:
:157 g/mol
:208-611-6
:2929
:GHS07
:535-15-9
:C₄H₆Cl₂O₂
:152…156 °C (1013 hPa)
:1.2904 g/cm³ (20 °C)
:60
:6.1 II