Formylhydrazine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide
CAS Number:624-84-0
:
Formula:CH₄N₂O
MDL Number:MFCD00007589
Boiling Pt:90 °C (0.7 mmHg)
Molecular Weight:60.06 g/mol
Melting Pt:150
:M-Clarity™ quality level = MQ100
EINECS:210-867-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together