Sodium butyrate
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: Butyric acid sodium salt, Sodium Butyrate
CAS Number:156-54-7
:
Formula:C₄H₇NaO₂
MDL Number:MFCD00002816
Density:1.324 g/cm³ (30 °C)
Molecular Weight:110.09 g/mol
Melting Pt:244
EINECS:205-857-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together