2-Bromobenzyl mercaptan ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Bromobenzylmercaptan, o-Bromotoluene-α-thiol
CAS Number:143888-85-1
:
Formula:C₇H₇BrS
MDL Number:MFCD04038925
Boiling Pt:256…257 °C
Density:1.527
Molecular Weight:203.1
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:2810
ADR:6.1 III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together