(2-Methoxyphenyl)acetonitrile ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Methoxybenzyl cyanide
CAS Number:7035-03-2
:
Formula:C₉H₉NO
MDL Number:MFCD00001902
Boiling Pt:141…143 °C (15 mmHg)
Molecular Weight:147.18 g/mol
Melting Pt:66…70 °C
Flash Pt:129 °C (264 °F)
Storage Temperature:Ambient
EINECS:230-314-5
UN:3439
ADR:6.1 III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together