4-Methoxybenzyl mercaptan ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Methoxy-α-toluenethiol, p-Methoxytoluene-α-thiol
CAS Number:6258-60-2
:
Formula:C₈H₁₀OS
MDL Number:MFCD00004871
Boiling Pt:90…95 °C (0.5 mmHg)
Density:1.112
Molecular Weight:154.23
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:228-393-6
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together