Order Entry
United Kingdom
ContactUsLinkComponent
 
3,5-Dimethylbenzyl bromide 98%
 
undefined
3,5-Dimethylbenzyl bromide 98%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Light yellow Low melting crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    36 to 40 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together