2'-Fluoro-4'-methoxyacetophenone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74457-86-6
:
Formula:C₉H₉FO₂
MDL Number:MFCD00042290
Boiling Pt:80…85 °C (.1 mmHg)
Molecular Weight:168.17
Melting Pt:50…54 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:50 to 54 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together