Kidney, nephron and kidney capsule
Catalog # 763-0124
Supplier: 3B Scientific
Specifications
- Description:Kidney, nephron and kidney capsule
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Three models on a common plate. Kidney in cross section enlarged 3×. Nephron enlarged 120× and kidney capsule enlarged 700×. Dimensions: 29×52×9 cm.