Coating Solution, βCoat®
Catalog # MSPPCOAT01
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pack type:Vial
- Pk:120 µl
- Source:
- Species:
- Description:Coating Solution, βCoat®, for the culture of EndoC-βH5®, EndoC-βH3®, EndoC-βH1® and ECN90® cells
- Environmentally Preferable:
Specifications
About this item
βCoat® is a coating solution specifically designed for the culture of EndoC-βH5®, EndoC-βH3®, EndoC-βH1® and ECN90® cells.
Each vial of βCoat® contains between 90µL and 130 µl of βCoat® reagent, depending on the batch’s concentration, and allows to prepare 10 ml of coating solution.
βCoat® is manufactured, including packaging and quality control, at Human Cell Design facility in compliance with Human Cell Design Quality Management System.
FOR RESEARCH USE ONLY.