Polyethylene glycol 300, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: PEG 300
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
Density:1.13 g/cm³ (20 °C)
Molecular Weight:300 g/mol
Melting Pt:–15…–10 °C
Flash Pt:220 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:500-038-2
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together