£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Enzymes
Proteinase K
Proteinase K
MOLEM70856393
:
Molekula
:
Proteinase K
MOLEM70856393
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 mg
CAS No.:
39450-01-6
Source:
Species:
Environmentally Preferable:
Storage conditions:
Freezer
Enzyme name:
Proteinase K