Cleanline Heavy Duty Drain Unblocker and Maintainer
BUNZ033359
: BUNZL
- Capacity:1 L
- Description:Cleanline heavy duty drain maintainer and unblocker
- Environmentally Preferable:
- Colour:Faint purple
- Density:1,2 - 1,25
- Formulation:Sodium hydroxide; caustic soda
- Pk:6
- pH value:>11,5
Cleanline heavy duty drain maintainer and unblocker will remove heavy organic residues from bathroom and kitchen drains.
- A fast-acting and powerful liquid drain cleaner
- Suitable for use in pipes, drains and U-bends
- Rapid results on blocked drains and pipework
A fast-acting and powerful liquid drain cleaner. Quickly removes fat, grease, hair, food blockages and other tenacious deposits. Rapid results on blocked drains and pipework from sinks, showers and industrial drainage systems. Suitable for use in pipes, drains and U-bends. Does not damage PVC or stainless steel. The one litre angled neck bottle allows users to easily apply the solution.
Store locked up. Store in a well-ventilated place. Keep cool.
: Causes severe skin burns and eye damage. Causes serious eye damage.