Order Entry
United Kingdom
ContactUsLinkComponent
Table clamp
Table clamp
Catalog # 241-7350
undefined
Table clamp
Catalog # 241-7350

Specifications

  • Spanning width:
    60 mm/ Ø13 mm
  • Description:
    Table clamp
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Aluminium alloy, powder coated.

  • For rods up to Ø 13 mm
  • For benches up to 60 mm
  • Hinged screws for rapid, easy fixing