Anti-TIN-Ag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-13712R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TIN-Ag
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:301-400/476
- Form:Liquid
- Antigen synonyms:TINAG|TIN1|TIN AG|TIN2|Tubulointerstitial nephritis antigen|TINAG_HUMAN|TIN-Ag
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:55 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TIN-Ag
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Mediates adhesion of proximal tubule epithelial cells via integrins alpha3-beta1 and alphaV-beta3. This is a non catalytic peptidase C1 family protein.
Type: Primary
Antigen: TIN-Ag
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 301-400/476
Host: Rabbit
Isotype: IgG
Reactivity: