- Antibody type:Primary
- Antigen name:PL6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Epitope:201-300/351
- Form:Liquid
- Antigen synonyms:TMEM115|Placental protein 6|transmembrane protein 115| PP6
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:38 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PL6
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Transmembrane proteins contain transmembrane domains that are usually characterized by alpha-helical structures. Transmembrane proteins exist as thermodynamically stable hetero- and homodimers that interact with the lipid bilayer and are involved in both material exchange and communication between the cell and the environment. PL6 is a 351 amino acid multi-pass membrane protein that is highly expressed in kidney and skeletal muscle with lower levels of expression detected in liver, placenta, prancreas, lung, heart and brain. PL6 contains one phosphoserine residue and several transmembrane domains, suggesting that it may participate in protein exchange and signaling events between cells.
Type: Primary
Antigen: PL6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/351
Host: Rabbit
Isotype: IgG
Reactivity: