Anti-ZA20D3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-13560R-A750
: Bioss
- Antibody type:Primary
- Antigen name:ZA20D3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:111-208/208
- Form:Liquid
- Antigen synonyms:fb11b11|DKFZp564F2116|Zinc finger A20 domain-containing protein 3|AWP1|ZA20D3|AN1-type zinc finger protein 6|Za20d3 zinc finger A20 domain containing 3|Associated with PRK1 protein|ZFAND5B|3110005P07Rik|ZFAN6_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:23 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ZA20D3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
ZA20D3 is a zinc-finger protein located on chromosome 15. It is associated with PRK1.
Type: Primary
Antigen: ZA20D3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 111-208/208
Host: Rabbit
Isotype: IgG
Reactivity: