Anti-FYCO1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-13237R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:FYCO1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1051-1150/1478
- Form:Liquid
- Gene ID:79443
- Antigen synonyms:CTRCT18|CATC2|FYVE and coiled-coil domain-containing protein 1|RUFY3|ZFYVE7|Zinc finger FYVE domain-containing protein 7|FYCO1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:167 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human FYCO1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
May mediate microtubule plus end-directed vesicle transport.
Type: Primary
Antigen: FYCO1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1051-1150/1478
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse