Anti-USP22 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-8611R-A750
: Bioss
- Antibody type:Primary
- Antigen name:USP22
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Xenopus laevis,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:121-220/525
- Form:Liquid
- Gene ID:23326
- Antigen synonyms:Ubiquitin-specific-processing protease 22|Ubiquitin carboxyl-terminal hydrolase 22|USP22|KIAA1063|Ubiquitin thioesterase 22|Deubiquitinating enzyme 22|USP3L
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:60 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human USP22
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.
Type: Primary
Antigen: USP22
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-220/525
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Xenopus laevis