Anti-TXNRD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-8299R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TXNRD1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:451-550/649
- Form:Liquid
- Gene ID:7296
- Antigen synonyms:Gene associated with retinoic and IFN-induced mortality 12 protein|Thioredoxin reductase 1, cytoplasmic|GRIM-12|GRIM12|TR1|TXNR|TRXR1|Gene associated with retinoic and interferon-induced mortality 12 protein|KDRF|Thioredoxin reductase TR1|TXNRD1|KM-12-derived reductase-like factor|TR
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:71 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TXNRD1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Isoform 1 may possess glutaredoxin activity as well as thioredoxin reductase activity and induces actin and tubulin polymerisation, leading to formation of cell membrane protrusions. Isoform 4 enhances the transcriptional activity of estrogen receptors alpha and beta while isoform 5 enhances the transcriptional activity of the beta receptor only. Isoform 5 also mediates cell death induced by a combination of interferon-beta and retinoic acid.
Type: Primary
Antigen: TXNRD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 451-550/649
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat