Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SGTA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-SGTA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-7830R-A750
 :  Bioss
Anti-SGTA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-7830R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    SGTA
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    181-280/313
  • Form:
    Liquid
  • Gene ID:
    6449
  • Antigen synonyms:
    AlphaSGT|Alpha-SGT|UBP|Small glutamine rich protein with tetratricopeptide repeats 1|Small glutamine-rich tetratricopeptide repeat-containing protein alpha|SGT|Sgt1|Small glutamine rich tetratricopeptide repeat TPR containing alpha|SGTA_HUMAN|Vpu-binding protein|Vpu binding protein|Stg|Small glutamine rich tetratricopeptide repeat-containing protein|SGTA|Alpha SGT|hSGT|Small glutamine rich tetratricopeptide repeat containing protein alpha
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    34 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SGTA
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Co-chaperone that binds directly to HSC70 and HSP70 and regulates their ATPase activity.

Type: Primary
Antigen: SGTA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 181-280/313
Host: Rabbit
Isotype: IgG
Reactivity: