Anti-BRD7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-7779R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BRD7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Epitope:401-500/651
- Form:Liquid
- Gene ID:29117
- Antigen synonyms:Bromodomain containing 7|Protein CELTIX-1|BRD7|75 kDa bromodomain protein|BRD7_HUMAN|CELTIX 1|CELTIX1|BRD 7|Bromodomain-containing protein 7|NAG4|BP75|bromodomain containing protein 7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:74 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BRD7
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a protein which is a member of the bromodomain-containing protein family. The product of this gene has been identified as a component of one form of the SWI/SNF chromatin remodeling complex, and as a protein which interacts with p53 and is required for p53-dependent oncogene-induced senescence which prevents tumor growth. Pseudogenes have been described on chromosomes 2, 3, 6, 13 and 14. Alternative splicing results in multiple transcript variants.
Type: Primary
Antigen: BRD7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-500/651
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse