Anti-Rab3A+Rab3B+Rab3C+Rab3D Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6180R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Rab3A+Rab3B+Rab3C+Rab3D
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:101-150/220
- Form:Liquid
- Antigen synonyms:Ras related protein Rab 3C|Ras related protein Rab 3D|Glioblastoma overexpressed|RAB3A|RAB3D member RAS oncogene family|GOV|RAD3D|D2 2|RAB16|Ras related protein Rab 3A|Ras related protein Rab 3B|RAB3B|RAB3C member RAS oncogene family|RAB3B member RAS oncogene family|RAB3C|RAB3A member RAS oncogene family
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:24 - 27 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Rab3A+Rab3B+Rab3C+Rab3D
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has four known isoforms: 3A, 3B, 3C and 3D. These proteins are expressed in the highest concentration in the brain and endocrine tissues and are involved in regulated exocytosis of hormones and neurotransmitters. Specifically Rab3's are thought to play a role in presynaptic vesicle trafficking and priming.
Type: Primary
Antigen: Rab3A+Rab3B+Rab3C+Rab3D
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-150/220
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat