Anti-AARE Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-5983R-A750
: Bioss
- Antibody type:Primary
- Antigen name:AARE
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:101-200/732
- Form:Liquid
- Gene ID:327
- Antigen synonyms:Acylamino acid releasing enzyme|D3F15S2|APEH|Acyl peptide hydrolase|AARE|D3S48E|Acylaminoacyl peptidase|APH|DNF15S2|N acylaminoacyl peptide hydrolase|ACPH_HUMAN|ACPH|Oxidized protein hydrolase|OPH
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:81 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human AARE
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
AARE (Acylamino-acid-releasing enzyme) is also known as Acyl-peptide hydrolase. It catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. It can play an important role in destroying oxidatively damaged proteins in living cells. Deletions of this gene locus corresponding to the protein are found in various types of carcinomas, including small cell lung carcinoma and renal cell carcinoma.
Type: Primary
Antigen: AARE
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/732
Host: Rabbit
Isotype: IgG
Reactivity: Human