Anti-DECR1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5056R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DECR1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:201-300/335
- Form:Liquid
- Gene ID:1666
- Antigen synonyms:RP23-338J24.3|4-enoyl-CoA reductase [NADPH]|2,4 dienoyl CoA reductase mitochondrial|SDR18C1|2,4 dienoyl CoA reductase 1|Short chain dehydrogenase/reductase family 18C member 1|4-dienoyl-CoA reductase [NADPH]|1200012F07Rik|DECR 1|2,4-dienoyl-CoA reductase, mitochondrial|DECR1|DECR|2|4 enoyl CoA reductase|Nadph|OTTMUSP00000004962|2,4 dienoyl CoA reductase [NADPH]|DECR_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:32 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DECR1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Auxiliary enzyme of beta-oxidation. It participates in the metabolism of unsaturated fatty enoyl-CoA esters having double bonds in both even- and odd-numbered positions. Catalyzes the NADP-dependent reduction of 2,4-dienoyl-CoA to yield trans-3-enoyl-CoA.
Type: Primary
Antigen: DECR1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/335
Host: Rabbit
Isotype: IgG
Reactivity: