Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Cytohesin 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-12922R-A750
 :  Bioss
Anti-Cytohesin 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-12922R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Cytohesin 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    21-120/400
  • Form:
    Liquid
  • Gene ID:
    9266
  • Antigen synonyms:
    PSCD2L| Arno|Sec7pL| ARNO protein|Sec7p-like| ARF nucleotide binding site opener|ARF exchange factor| CLM2|Sec7B| Cytohesin 2|SEC7 homolog B| PSCD2L, formerly|Sec7p L| Cyth2|SEC7L| CTS18.1| MGC80440| PH, SEC7 and coiled-coil domain-containing protein 2| Pleckstrin homology Sec7 and coiled coil domains 2| Pleckstrin homology Sec7 and coiled coil domains protein 2|Sec7| MGC137537| CTS18|CYH2_HUMAN| PSCD2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    47 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Cytohesin 2
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

The ADP-ribosylation factor (Arf) family comprises a group of structurally and functionally conserved 21 kDa proteins, which are members of the Ras superfamily of regulatory GTP-binding proteins. Arf is involved in intracellular protein traffic to and within the Golgi complex. Arf has a number of disparate activities including maintenance of organelle integrity, assembly of coat proteins, as a co-factor for cholera toxin and as an activator of phospholipase D. Like other small GTPases, Arf is found to be active when bound to GTP and inactive when bound to GDP. Arf's activation is dependent upon guanine nucleotide exchange factors (GEFs) which increase the rate of exchange of bound GDP with GTP. All GEFs have a highly conserved Sec7 domain. GEF activity lies in the Sec7 domain and this activity has been shown to be inhibited by the fungal metabolite brefeldin-A (BFA). A small group of GEFs which are insensitive to brefeldin-A (BFA) include cytohesin-1 (B2-1), cytohesin-2 (ARNO), cytohesin-3 (ARNO3), and cytohesin-4. All cytohesins function in the cell periphery and contain a pleckstrin homology (PH) domain. The PH domain has been shown to interact with phosphatidylinositol 3,4,5-triphosphate and is believed to promote membrane targeting of the cytohesins. Recruitment of the cytohesins to the membranes can occur in response to tyrosine kinase receptor activation. This response appears to require the activation of phosphatidylinositol 3-kinase (PI 3-kinase).

Type: Primary
Antigen: Cytohesin 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/400
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat