Anti-APC1 Ser688 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-12482R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:APC1 Ser688
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Environmentally Preferable:
- Form:Liquid
- Gene ID:64682
- Antigen synonyms:Mitotic checkpoint regulator|TSG 24|Testis-specic gene 24 protein|Anaphase-promoting complex subunit 1|Protein Tsg24|Cyclosome subunit 1|Apc1 phospho S688|Protein Tsg 24|APC1_HUMAN|Apc 1|anaphase-promoting complex 1 meiotic checkpoint regulator|Meiotic checkpoint regulator|ANAPC1Ser688|TSG24|ANAPC 1|APC1|Anaphase promoting complex subunit 1|ANAPC1|MCPR
- Modification:Phosphorylation (Ser688)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:215 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human APC1 around the phosphorylation site of Ser688
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a subunit of the anaphase-promoting complex. This complex is an E3 ubiquitin ligase that regulates progression through the metaphase to anaphase portion of the cell cycle by ubiquitinating proteins which targets them for degradation.
Type: Primary
Antigen: APC1 Ser688
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Rat