Specifications
- Antibody type:Primary
- Antigen name:alpha B Crystallin Ser59
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Environmentally Preferable:
- Form:Liquid
- Gene ID:1410
- Antigen synonyms:Crystallin alpha polypeptide 2|AlphaB-crystallin|alpha B Crystallin phospho S59|HSPB5|Heat shock 20 kD like protein|Alpha-crystallin B chain|NY REN 27 antigen|Crystallin alpha B|CRYA2|Renal carcinoma antigen NY REN 27|p-alpha B Crystallin S59|alpha B Crystallin phospho Ser59|Heat shock protein beta-5|CRYAB_HUMAN|Rosenthal fiber component|CTPP2|p-alpha B Crystallin Ser59|CTPP 2|Renal carcinoma antigen NY-REN-27|AACRYA|Alpha B crystallin|Heat shock protein beta 5|Alpha crystallin B chain|AlphaB crystallin|CRYAB
- Modification:Phosphorylation (Ser59)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human alpha B Crystallin around the phosphorylation site of Ser59
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Alpha crystallins are composed of two gene products: alpha-A and alpha-B, for acidic and basic, respectively. Alpha crystallins can be induced by heat shock and are members of the small heat shock protein (sHSP also known as the HSP20) family. They act as molecular chaperones although they do not renature proteins and release them in the fashion of a true chaperone; instead they hold them in large soluble aggregates. Post-translational modifications decrease the ability to chaperone. These heterogeneous aggregates consist of 30-40 subunits; the alpha-A and alpha-B subunits have a 3:1 ratio, respectively. Two additional functions of alpha crystallins are an autokinase activity and participation in the intracellular architecture. Alpha-A and alpha-B gene products are differentially expressed; alpha-A is preferentially restricted to the lens and alpha-B is expressed widely in many tissues and organs. Elevated expression of alpha-B crystallin occurs in many neurological diseases; a missense mutation cosegregated in a family with a desmin-related myopathy.
Type: Primary
Antigen: alpha B Crystallin Ser59
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse