Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Peroxin 5R Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-12096R-A680
 :  Bioss
Anti-Peroxin 5R Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-12096R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Peroxin 5R
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Environmentally Preferable:
  • Epitope:
    101-200/626
  • Form:
    Liquid
  • Antigen synonyms:
    Peroxisome biogenesis factor 5 like|PEX5R|Pex5Rp|PEX5R_HUMAN|PXR2B|PEX5-like protein|Peroxin 5 related protein|PEX2 related protein
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    70 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Peroxin 5R/PEX5-related protein
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Peroxisomes are single-membrane bound organelles present in virtually all eukaryotic cells. They are involved in numerous catabolic and anabolic pathways, including beta-oxidation of very long chain fatty acids, metabolism of hydrogen peroxide, plasmalogen biosynthesis and bile acid synthesis. The Peroxin gene family, which includes more than 20 members, is required for peroxisome biogenesis. Peroxin 5R, also known as PEX5-related protein or Peroxisome biogenesis factor 5-like, is a 626 amino acid protein that is mainly expressed in brain, with some expression in testis and pancreas. Peroxin 5R contains five TPR repeats, which enable protein-protein interactions and assembly of large multiprotein complexes. There are three isoforms of Peroxin 5R that are produced as a result of alternative splicing events. These isoforms bind C-terminal peroxisome-targeting signals in a similar manner to Peroxin-5. Peroxin 5R interacts with Rab 8b, possibly playing a role in vesicular trafficking and neurotransmitter release.

Type: Primary
Antigen: Peroxin 5R
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/626
Host: Rabbit
Isotype: IgG
Reactivity: